missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, For Lead, 5% (w/v), LabChem™

Catalog No. LC249401
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC249401 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC249401 Supplier LabChem Supplier No. LC249401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-83-7 , 7732-18-5
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
CAS Max % 95, 5
Linear Formula Na2SO3
Solubility Information Soluble in water
Formula Weight 126.04
Percent Purity 5% (w/v)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Sulfite, For Lead
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature