missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfite, ≥98% (RT), MilliporeSigma™ Supelco™

Catalog No. 111015510
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015510 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015510 Supplier Merck Emd Millipore Supplier No. 089811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7757-83-7
Linear Formula Na2 SO3
MDL Number MFCD00003503
Synonym E221
Solubility Soluble in water
Type Analytical Standard
Quantity 1 g
Formula Weight 126.04
Percent Purity ≥98% (RT)
Grade Analytical Standard