missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, 2.5% (w/v), LabChem™

Catalog No. LC249352
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC249352 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC249352 Supplier LabChem Supplier No. LC249352

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-83-7 , 7732-18-5
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
CAS Max % 97.5, 2.5
Physical Form Liquid
Quantity 1 L
Linear Formula Na2SO3
Solubility Information Soluble in water
Formula Weight 126.04
Percent Purity 2.5% (w/v)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds
Chemical Name or Material Sodium Sulfite
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature