missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, 2.5% (w/v), Ricca Chemical

Catalog No. R75790001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R75790001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R75790001A Supplier Ricca Chemical Company Supplier No. R75790001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-83-7 , 7732-18-5
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
CAS Min % 96.0, 2.39
CAS Max % 100.0, 2.49
Percent Purity 2.5%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 9 to 9.5
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 2.5% (w/v)
Chemical Name or Material Sodium Sulfite
Show More Show Less