missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfite, 10% (w/v), Ricca Chemical
$138.81 - $278.00
Chemical Identifiers
| CAS | 7757-83-7 |
|---|---|
| Molecular Formula | Na2O3S |
| Molecular Weight (g/mol) | 126.037 |
| InChI Key | GEHJYWRUCIMESM-UHFFFAOYSA-L |
| Synonym | sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit |
| PubChem CID | 24437 |
| ChEBI | CHEBI:86477 |
| IUPAC Name | disodium;sulfite |
| SMILES | [O-]S(=O)[O-].[Na+].[Na+] |
Chemical Identifiers
| 7757-83-7 | |
| 126.037 | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| CHEBI:86477 | |
| [O-]S(=O)[O-].[Na+].[Na+] |
| Na2O3S | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| 24437 | |
| disodium;sulfite |
Specifications
| Colorless | |
| 89.0,8.91 | |
| Na2O3S | |
| GEHJYWRUCIMESM-UHFFFAOYSA-L | |
| disodium;sulfite | |
| 24437 | |
| 10% | |
| Natural Poly Bottle | |
| 9 to 9.5 | |
| Liquid | |
| Sodium Sulfite |
| 7757-83-7,7732-18-5 | |
| 93.0,9.28 | |
| sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
| [O-]S(=O)[O-].[Na+].[Na+] | |
| 126.037 | |
| CHEBI:86477 | |
| Laboratory | |
| Aqueous Solution | |
| 500 mL | |
| 10% (w/v) |