missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfite, 10% (w/v), Ricca Chemical

Catalog No. 758516
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
758516 500 mL Natural Poly Bottle
758532 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 758516 Supplier Ricca Chemical Company Supplier No. 758516

Chemical Identifiers

CAS 7757-83-7 , 7732-18-5
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 9 to 9.5
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Sodium Sulfite
Show More Show Less