missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide, Solution No. 1 for Zinc, Certified, 3% (w/v), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 97, 3 |
| Linear Formula | Na2S·9H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 240.17 |
| Percent Purity | 3% (w/v) |
| Grade | Certified |
| Identification | Passes Test |
| Odor | Sulfide |
| Packaging | Poly Bottle |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye damage. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Collect spillage. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |