missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide, 5% (w/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 93.0, 4.67 |
| CAS Max % | 97.0, 4.86 |
| Percent Purity | 5% |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Solvent or Matrix | Aqueous Solution |
| pH | Alkaline |
| Quantity | 1 L |
| Physical Form | Liquid |
| Show More |