missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide, 3% (w/v), Ricca Chemical
$82.31 - $304.75
Chemical Identifiers
| CAS | 1313-84-4 |
|---|---|
| Molecular Formula | H18Na2O9S |
| Molecular Weight (g/mol) | 240.17 |
| MDL Number | MFCD00149184 |
| InChI Key | ZGHLCBJZQLNUAZ-UHFFFAOYSA-N |
| IUPAC Name | disodium nonahydrate sulfanediide |
| SMILES | O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
75704
|
Ricca Chemical Company
75704 |
120 mL | Amber Glass Bottle |
Each for $82.31
|
|
||||
|
7570-16
|
Ricca Chemical Company
757016 |
500 mL | Amber Glass Bottle |
Each for $202.06
|
|
||||
|
7570-32
|
Ricca Chemical Company
757032 |
1 L | Amber Glass Bottle |
Each for $304.75
|
|
||||
Chemical Identifiers
| 1313-84-4 | |
| 240.17 | |
| ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
| O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
| H18Na2O9S | |
| MFCD00149184 | |
| disodium nonahydrate sulfanediide |
Specifications
| Colorless | |
| 95.0,2.86 | |
| H18Na2O9S | |
| ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
| disodium nonahydrate sulfanediide | |
| 3% | |
| Amber Glass Bottle | |
| Alkaline | |
| Liquid | |
| Sodium Sulfide |
| 1313-84-4,7732-18-5 | |
| 99.0,2.97 | |
| MFCD00149184 | |
| O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] | |
| 240.17 | |
| Laboratory | |
| Aqueous Solution | |
| 120 mL | |
| 3% (w/v) |