missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide, 3% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 757016
Specifications
| Sodium Sulfide | |
| 1313-84-4,7732-18-5 | |
| 99.0,2.97 | |
| MFCD00149184 | |
| O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] | |
| 240.17 | |
| Laboratory | |
| Aqueous Solution | |
| 500 mL | |
| 3% (w/v) |
| Colorless | |
| 95.0,2.86 | |
| H18Na2O9S | |
| ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
| disodium nonahydrate sulfanediide | |
| 3% | |
| Amber Glass Bottle | |
| Alkaline | |
| Liquid |
Chemical Identifiers
| 1313-84-4 | |
| 240.17 | |
| ZGHLCBJZQLNUAZ-UHFFFAOYSA-N | |
| O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--] |
| H18Na2O9S | |
| MFCD00149184 | |
| disodium nonahydrate sulfanediide |