missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide, 3% (w/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 95.0, 2.86 |
| CAS Max % | 99.0, 2.97 |
| Percent Purity | 3% |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Solvent or Matrix | Aqueous Solution |
| pH | Alkaline |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Show More |