missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium sulfate decahydrate, 99%, for biochemistry

Catalog No. AC447935000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447935000 500 g Plastic bottle
AC447930025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447935000 Supplier Thermo Scientific Chemicals Supplier No. 447935000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7727-73-3 | Na2O4S·10H2O | 322.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7727-73-3
Molecular Formula Na2O4S·10H2O
Molecular Weight (g/mol) 322.2
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 884.0°C
Quantity 500 g
Packaging Plastic bottle
Assay Percent Range 99%
Linear Formula Na2SO4·10H2O
Solubility Information Clear colorless
Formula Weight 322.2
Percent Purity 99%
Grade Biochemical
Chemical Name or Material Sodium sulfate decahydrate

RUO – Research Use Only