missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™

Catalog No. S41510S
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
50 kg
200 lb.
Packaging:
Fiber Drum
Glass Bottle
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S41510S 10 kg Poly Pail
S415500 500 g Glass Bottle
S415200LB 200 lb. Fiber Drum
S4151 1 kg Poly Bottle
S415212 2.5 kg Glass Bottle
S41550 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S41510S Supplier Thermo Fisher Scientific Supplier No. S415 10
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color White
pH 5 to 8
Physical Form Solid
Quantity 10 kg
Assay Percent Range ≥99.0 %
Formula Weight 142.04
Percent Purity ≥99.0%
Grade Suitable for pesticide residue analysis
Identification Pass Test
Loss on Ignition 0.5% max.
Packaging Poly Pail
Decomposition Information Decomposition Information: >890°C
Chemical Name or Material Sodium Sulfate Anhydrous
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category SIGNAL WORD: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0