missing translation for 'onlineSavingsMsg'
Learn More

Sodium Stearyl Fumarate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014152
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014152 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014152 Supplier Merck Emd Millipore Supplier No. PHR14441G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Sodium Stearyl Fumarate is a water soluble lubricant, which is widely used in the pharmaceutical industry for the compression of tablets. Its ability to reduce friction, adhesion to the punches, tablet strength and the disintegration time was investigated.

Specifications

CAS 4070-80-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C22H39 NaO4
Synonym Sodium stearyl fumarate; (E)-2-Butenedioic acid monooctadecyl ester sodium salt; Sodium octadecyl fumarate
Type Certified Reference Material
Quantity 1 g
Formula Weight 390.53
Grade Certified Reference Material