missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Catalog No. PLNA22Y
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLNA22Y 125 mL
PLNA22X 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLNA22Y Supplier SPEX CertiPrep Group LLC Supplier No. PLNA22Y

missing translation for 'validMainframeMsg'

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Na1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Sodium Standard
Packaging Triple-leached LDPE bottle
CAS 7440-23-5, 7732-18-5, 7697-37-2
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula Na
Technique AA, ICP
MDL Number MFCD00085307
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES [Na]
Solvent or Matrix 2% HNO3
IUPAC Name sodium
Molecular Weight (g/mol) 22.99
Quantity 125 mL
PubChem CID 5360545
ChEBI CHEBI:26708
Grade Assurance
Show More Show Less

Product intended for laboratory use only