missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Standard, 10 ppm Na, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 10ppm Na |
| CAS Min % | 98.0, 0.0025 |
| CAS Max % | 100.0, 0.0026 |
| Equivalence | 1 mL = 0.01 mg Na |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Quantity | 1 L |
| Chemical Name or Material | Sodium Standard |