missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Standard, 10 ppm Na, Ricca Chemical
Supplier: Ricca Chemical Company 75951
Specifications
| Sodium Standard | |
| 7647-14-5 , 7732-18-5 | |
| 10ppm Na | |
| 1 mL = 0.01 mg Na | |
| ClNa | |
| Colorless | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 4 L | |
| CHEBI:26710 |
| Natural Poly Bottle | |
| 98.0,0.0025 | |
| 100.0,0.0026 | |
| Liquid | |
| MFCD00003477 | |
| 100°C | |
| [Na+].[Cl-] | |
| 58.44 | |
| 5234 |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |