missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™

Catalog No. ORGNA82Z
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ORGNA82Z 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ORGNA82Z Supplier SPEX CertiPrep Group LLC Supplier No. ORGNA82Z

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Na1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-23-5 , 8042-47-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.5, 99.5
CAS Max % 0.5, 99.5
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Base Oil 20
Industry Type Petrochemical Testing
Quantity 50 g
Grade Oil/Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Base Oil 1000μg/g ORG-NA8-2Z
Base Oil 5000μg/g ORG-NA8-4Z
Chemical Name or Material Sodium Oil Standard
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only