missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenate, Purum p.a., ≥98.0% (T), Solstice

Catalog No. 6001550
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001550 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001550 Supplier Solstice Supplier No. 71948500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
Synonym sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
PubChem CID 25960
ChEBI CHEBI:77775
IUPAC Name disodium;selenate
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 500 g
Linear Formula Na2SeO4
UN Number UN2630
Formula Weight 188.94g/mol
Percent Purity ≥98% (T)
Grade Not Available
Packaging Poly bottle
Chemical Name or Material Sodium selenate
Hazard Statement P261-P273-P301 + P310-P311-P501
RTECSNumber VS6650000