missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate Anhydrous, Technical (Tetrasodium Pyrophosphate), Reagents

Catalog No. 01336981
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336981 10 kg
01336980 2.5 kg
01336979 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336981 Supplier Labchem Inc Supplier No. C232458010KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Pyrophosphate Anhydrous, Technical (Tetrasodium Pyrophosphate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7722-88-5
Molecular Formula Na4O7P2
Molecular Weight (g/mol) 265.90
InChI Key FQENQNTWSFEDLI-UHFFFAOYSA-J
Synonym Tetrasodium Pyrophosphate
IUPAC Name tetrasodium (phosphonatooxy)phosphonate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Melting Point 880°C
Color Colorless/White
Physical Form Solid
Quantity 10 kg
Grade Reagent
Packaging Poly Pail
Specific Gravity 2.53
Chemical Name or Material Sodium Pyrophosphate Anhydrous, Technical (Tetrasodium Pyrophosphate)