missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate, 10-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Supplier:  Avantor 385005

 View more versions of this product

Catalog No. 02004227

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Sodium Pyrophosphate
13472-36-1
2.5 kg
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
446.05
446.06
Poly Bottle
10-Hydrate
Crystals
H20Na4O17P2
VZWGHDYJGOMEKT-UHFFFAOYSA-J
tetrasodium;phosphonato phosphate;decahydrate
3084150
BAKER ANALYZED™ ACS Reagent
1L = 1.82kg

Chemical Identifiers

13472-36-1
446.05
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
tetrasodium;phosphonato phosphate;decahydrate
H20Na4O17P2
VZWGHDYJGOMEKT-UHFFFAOYSA-J
3084150
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions