missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, 50% (w/v), LabChem™
Supplier: LabChem LC223102
Specifications
| Sodium Potassium Tartrate | |
| For Nitrogen (Ammonia), Nitrogen (Organic) | |
| 50,50 | |
| NaKC4H4O6·4H2O | |
| Soluble in water | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
| 282.218 | |
| 282.23 | |
| Certified | |
| Poly Bottle | |
| Carbon monoxide; Carbon dioxide | |
| Liquid |
| Colorless | |
| 6381-59-5 , 7732-18-5 | |
| C4H12KNaO10 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
| 2724148 | |
| 50% (w/v) | |
| Passes Test | |
| Passes Test | |
| 1 L |
Chemical Identifiers
| 6381-59-5 | |
| 282.218 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
| C4H12KNaO10 | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| 2724148 | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Refrigerate