missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, 50% (w/v), LabChem™

Catalog No. p-5388598
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC223107 125 mL
LC223101 500 mL
LC223102 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC223107 Supplier LabChem Supplier No. LC223107
Only null left

Chemical Identifiers

CAS 6381-59-5 , 7732-18-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.218
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]

Specifications

Color Colorless
CAS Max % 50
Linear Formula NaKC4H4O6·4H2O
Solubility Information Soluble in water
Formula Weight 282.23
Percent Purity 50% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Suitability Passes Test
Decomposition Information Carbon monoxide; Carbon dioxide
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Refrigerate