missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Monobasic, Monohydrate, Multi-Compendial, U.S.P., J.T. Baker™
Specifications
Specifications
| Quantity | 500 g |
| Formula Weight | 137.99 |
| Percent Purity | 98% |
| Grade | Multi-Compendial/USP |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.04kg |
| Chemical Name or Material | Sodium Phosphate, Monobasic, Monohydrate |