missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Phosphate, Monobasic, Monohydrate, Molecular biology grade, Calbiochem™,
Chemical Identifiers
| CAS | 10049-21-5 |
|---|---|
| Molecular Formula | H5NaO5P |
| Molecular Weight (g/mol) | 139.00 |
| MDL Number | MFCD00149208 |
| InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
| Synonym | NaH2PO4 |
| PubChem CID | 516949 |
| IUPAC Name | sodium phosphoric acid hydrate |
| SMILES | O.[Na+].OP(O)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
567549250GM
|
MilliporeSigma™
567549250GM |
250 g | Glass Bottle |
N/A
|
N/A | ||||
|
5675491KG
|
MilliporeSigma™
5675491KG |
1 kg | Glass Bottle |
N/A
|
N/A | ||||
Chemical Identifiers
| 10049-21-5 | |
| 139.00 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| 516949 | |
| O.[Na+].OP(O)(O)=O |
| H5NaO5P | |
| MFCD00149208 | |
| NaH2PO4 | |
| sodium phosphoric acid hydrate |
Specifications
| White | |
| H5NaO5P | |
| NaH2PO4 | |
| O.[Na+].OP(O)(O)=O | |
| 139.00 | |
| ≥99% (Titration) |
| 10049-21-5 | |
| MFCD00149208 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium phosphoric acid hydrate | |
| 516949 | |
| Molecular Biology |