missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Phosphate, Monobasic, Monohydrate, Molecular biology grade, Calbiochem™,

Supplier:  MilliporeSigma™ 5675491KG

Quantity 1 kg
Packaging Glass Bottle

 View more versions of this product

Catalog No. 5675491KG


Only null left

Description

Description

Specifications

Chemical Identifiers

10049-21-5
139.00
BBMHARZCALWXSL-UHFFFAOYSA-N
516949
O.[Na+].OP(O)(O)=O
H5NaO5P
MFCD00149208
NaH2PO4
sodium phosphoric acid hydrate

Specifications

White
1 kg
H5NaO5P
NaH2PO4
O.[Na+].OP(O)(O)=O
139.00
≥99% (Titration)
10049-21-5
Glass Bottle
MFCD00149208
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium phosphoric acid hydrate
516949
Molecular Biology
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions