missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic Monohydrate (Cryst. and Granular/USP/FCC), Fisher Chemical

Catalog No. S468500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S468500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S468500 Supplier Thermo Fisher Scientific Supplier No. S468 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Melting Point 100°C
Color White
pH 4.1 to 4.5
Physical Form Solid
Quantity 500 g
Formula Weight 137.99
Percent Purity 98.0 to 103.0% (FCC)
Grade FCC/USP
Identification Pass Test (USP/FCC), Pass Test (FCC), Pass Test (USP)
Loss on Drying 10.0 to 15.0% (FCC)
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate Monobasic Monohydrate
Show More Show Less