missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
| Melting Point | 100.0°C |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 250 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 99.5% |
| Linear Formula | NaH2PO4·H2O |
| Merck Index | 15,879 |
| Solubility Information | Clear colorless |
| Formula Weight | 137.99 |
| Percent Purity | 99.5% |
| Show More |
RUO – Research Use Only