missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Endotoxin Tested, Monobasic, Monohydrate, Crystal, U.S.P. - F.C.C., J.T. Baker™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 500 g |
| Formula Weight | 137.99 |
| Percent Purity | 98.0% to 103.0% |
| Grade | FCC/USP |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.04kg |
| Chemical Name or Material | Sodium Phosphate, Endotoxin Tested, Monobasic, Monohydrate |