missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Phosphate, Dibasic, Molecular biology grade, Calbiochem™,
$80.20 - $256.00
Chemical Identifiers
| CAS | 7558-79-4 |
|---|---|
| Molecular Formula | HNa2O4P |
| Molecular Weight (g/mol) | 141.96 |
| MDL Number | MFCD00003496 |
| InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
| Synonym | Na2HPO4 |
| PubChem CID | 24203 |
| ChEBI | CHEBI:34683 |
| IUPAC Name | disodium hydrogen phosphate |
| SMILES | [Na+].[Na+].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
567547250GM
|
MilliporeSigma™
567547250GM |
250 g | Glass Bottle |
Each for $80.20
|
|
||||
|
5675471KG
|
MilliporeSigma™
5675471KG |
1 kg | Glass Bottle |
Each for $256.00
|
|
||||
Chemical Identifiers
| HNa2O4P | |
| MFCD00003496 | |
| Na2HPO4 | |
| CHEBI:34683 | |
| [Na+].[Na+].OP([O-])([O-])=O |
Specifications
| White | |
| HNa2O4P | |
| Na2HPO4 | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| CHEBI:34683 | |
| Molecular Biology |
| 7558-79-4 | |
| MFCD00003496 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium hydrogen phosphate | |
| 24203 | |
| ≥99% (Titration) |