missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Phosphate, Dibasic, Molecular biology grade, Calbiochem™,

Catalog No. 567547250GM
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
567547250GM 250 g Glass Bottle
5675471KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 567547250GM Supplier MilliporeSigma™ Supplier No. 567547250GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym Na2HPO4
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
Quantity 250 g
Packaging Glass Bottle
Percent Purity ≥99% (Titration)
Grade Molecular Biology