missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Phosphate, Dibasic, Molecular biology grade, Calbiochem™,

Supplier:  MilliporeSigma™ 5675471KG

Quantity 1 kg
Packaging Glass Bottle

 View more versions of this product

Catalog No. 5675471KG


Only null left

Description

Description

Specifications

Chemical Identifiers

7558-79-4
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
disodium hydrogen phosphate
HNa2O4P
MFCD00003496
Na2HPO4
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O

Specifications

White
1 kg
HNa2O4P
Na2HPO4
[Na+].[Na+].OP([O-])([O-])=O
141.96
CHEBI:34683
Molecular Biology
7558-79-4
Glass Bottle
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
24203
≥99% (Titration)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions