missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Dibasic Dihydrate (USP/FCC/EP/BP), Fisher Chemical™
Specifications
Specifications
| Color | White |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Identification | Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP/BP) |
| Loss on Drying | 18.0 to 22.0% (FCC), 18.5 to 21.5% (USP), 18.5 to 21.5% (USP), 18.0 to 22.0% (FCC), 19.5 to 21.0% (EP/BP), 19.5 to 21.0% (BP), 19.5 to 21.0% (EP) |
| Packaging | Poly Bottle |
| Linear Formula | Na2HPO4·2H2O |
| Formula Weight | 177.99 |
| Percent Purity | ≥98.0% (FCC) |
| Grade | BP/EP/FCC/USP |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview Product dust may be irritating to eyes, skin and respiratory system. Wear personal protective equipment. Ensure adequate ventilation. Use personal protective equipment. Avoid contact with skin, eyes and clothing. Do not breathe dust. Rinse with water. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. NFPA Health:0 Flammability:0 Instability:0 |