missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, Anhydrous, U.S.P. - F.C.C., J.T. Baker™
Specifications
Specifications
| Quantity | 1 kg |
| Formula Weight | 141.96 |
| Percent Purity | 98.0% to 100.5% |
| Grade | FCC/USP |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 2.07kg |
| Chemical Name or Material | Sodium Phosphate, Dibasic, Anhydrous |