missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phenate Solution, for Ammonia Nitrogen Analysis, Ricca Chemical

For Analysis of Ammonia Nitrogen

$188.34 - $285.03

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7518-16
View Documents
Ricca Chemical Company
751816
500 mL Natural Poly Bottle
Each for $188.34
Only null left
 
7518-32
SDS View Documents
Ricca Chemical Company
751832
1 L Natural Poly Bottle
Each for $285.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
dihydrogen oxide, dihydrogen monoxide

Specifications

Colorless to Tan
87.0,8.54,2.78
dihydrogen oxide, dihydrogen monoxide
Natural Poly Bottle
500 mL
Sodium Phenate Solution
7732-18-5,108-95-2,1310-73-2
90.0,8.89,2.89
Laboratory
∼10.8
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'