missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phenate Solution, for Ammonia Nitrogen Analysis, Ricca Chemical
For Analysis of Ammonia Nitrogen
Supplier: Ricca Chemical Company 751832
Specifications
| Sodium Phenate Solution | |
| 7732-18-5,108-95-2,1310-73-2 | |
| 90.0,8.89,2.89 | |
| Laboratory | |
| ∼10.8 | |
| Liquid |
| Colorless to Tan | |
| 87.0,8.54,2.78 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Natural Poly Bottle | |
| 1 L |
Chemical Identifiers
| 7732-18-5 | |
| dihydrogen oxide, dihydrogen monoxide |