missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate, 0.60 M, Ricca Chemical

Catalog No. R751400010F
Change view
Click to view available options
Quantity:
10 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R751400010F 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R751400010F Supplier Ricca Chemical Company Supplier No. R751400010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7775-27-1 , 7732-18-5
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Assay Percent Range 0.600 ± 0.003 M at 20.0°C
Odor Odorless
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 10 L
Chemical Name or Material Sodium Persulfate, 0.6M, TOC Reagent