Learn More
Sodium peroxide, 95%
Description
Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.
Solubility
Soluble in acid. Insoluble in alkali.
Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. It reacts with water violently to give sodium hydroxide and hydrogen peroxide. Incompatible with strong reducing agents, organic materials, alcohols and powdered metals.
Specifications
Specifications
| Melting Point | 460°C (decomposition) |
| Physical Form | Beads |
| Quantity | 250 g |
| UN Number | UN1504 |
| Merck Index | 14,8655 |
| Solubility Information | Soluble in acid. Insoluble in alkali. |
| Formula Weight | 77.98 |
| Percent Purity | 95% |
| Grade | Ungraded |
| Sensitivity | Air sensitive; Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H271-H290-H314-H335 |
| Hazard Statement | GHS H Statement H271-H314 May cause fire or explosion; strong oxidizer. Causes severe skin burns and eye damage. |
| Precautionary Statement | P210-P220-P221-P234-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P390-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: SODIUM PEROXIDE |
| EINECSNumber | 215-209-4 |
| RTECSNumber | WD3450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only