missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxide, 95%

Catalog No. AAL1330618
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1330618 50 g
AAL1330630 250 g
AAL133060B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1330618 Supplier Thermo Scientific Chemicals Supplier No. L1330618

CAS: 1313-60-6 | Na2O2 | 77.978 g/mol

Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

Solubility
Soluble in acid. Insoluble in alkali.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. It reacts with water violently to give sodium hydroxide and hydrogen peroxide. Incompatible with strong reducing agents, organic materials, alcohols and powdered metals.

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Melting Point 460°C (decomposition)
Quantity 50 g
UN Number UN1504
Merck Index 14,8655
Solubility Information Soluble in acid. Insoluble in alkali.
Formula Weight 77.98
Percent Purity 95%
Sensitivity Air sensitive; Hygroscopic
Density 2.805
Chemical Name or Material Sodium peroxide
Hazard Category H271-H290-H314-H335
Hazard Statement GHS H Statement
H271-H314
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P220-P221-P234-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: SODIUM PEROXIDE
EINECSNumber 215-209-4
RTECSNumber WD3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only