missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate Electrolyte, Saturated Solution in Glacial Acetic Acid, Ricca Chemical

Catalog No. p-3252395
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
75054 120 mL Amber Glass Bottle
750516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75054 Supplier Ricca Chemical Company Supplier No. 75054
Only null left

Chemical Identifiers

CAS 7601-89-0 , 64-19-7
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.436
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]

Specifications

Color Colorless
CAS Min % 89.0, 8.85
CAS Max % 93.0, 9.21
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Glacial Acetic Acid
pH <2
Quantity 120 mL
Physical Form Liquid With Precipitate
Chemical Name or Material Sodium Perchlorate Electrolyte, Saturated Solution