accessibility menu, dialog, popup

UserName

CAS RN 7601-89-0

NaOClOOO

CAS RN 7601-89-0

IUPAC Name: sodium perchlorate
Synonyms: sodium perchlorate
Molecular Weight (g/mol): 122.44
Molecular Formula: ClNaO4
InChi Key: BAZAXWOYCMUHIX-UHFFFAOYSA-M
SMILES: [Na+].[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (14)
Grade 
  • (6)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (6)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (6)
  • (10)
  • (6)
  • (14)
  • (6)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)