missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium p-Styrenesulfonate Hydrate 93.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 100 g |
| Formula Weight | 206.19 |
| Percent Purity | ≥93.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium p-Styrenesulfonate Hydrate |
Safety and Handling
| EINECSNumber | (3)-1903&(3)-1948 |
| TSCA | Yes |