missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxide

Numéro de catalogue. AA3671214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA3671214 25 g
AA3671222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA3671214 Fournisseur Thermo Scientific Chemicals Code fournisseur 03671214
Il en reste null

CAS: 1313-59-3 | Na2O | 61.979 g/mol

Sodium oxide is used in chemical manufacturing, ceramics and glasses. Ethylbenzene dehydrogenation and carbon dioxide shift-reaction occurs using a sodium oxide/alumina catalyst. It is employed as catalyzing gasification of CO2 on carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 1313-59-3
Molecular Formula Na2O
Molecular Weight (g/mol) 61.979
MDL Number MFCD00046201
InChI Key KKCBUQHMOMHUOY-UHFFFAOYSA-N
Synonym sodium oxide, disodium oxide, disodium monoxide, disodium oxygen-2 anion, disodium oxidandiide, sodium oxide 25g, disodium oxygen 2-, 2na.o
PubChem CID 73971
IUPAC Name disodium;oxygen(2-)
SMILES [O-2].[Na+].[Na+]

Spécifications

Melting Point >400°C (decomposition)
Physical Form Beads
Quantity 25 g
UN Number UN1825
Merck Index 14,8651
Solubility Information Soluble in water.
Formula Weight 61.98
Sensitivity Air and moisture sensitive
Density 2.27 g/mL
Chemical Name or Material Sodium oxide
Hazard Category H271-H314-H335
Hazard Statement GHS H Statement
H271-H314
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: I
EINECSNumber 215-208-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only