Learn More
Sodium orthovanadate, 100mM aq. soln.
Description
Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium orthovanadate, is used as an inhibitor of several phosphatases and kinases such as protein tyrosine phosphatases (PTPs), alkaline phosphatase (ALP), and ATPase. It also acts as an inhibitor of adenosine kinase (AK) and phosphofructokinase (PFK). It also suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.
Solubility
It is soluble in water (50 mg/ml, )
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Linear Formula | Na3VO4 |
| Solubility Information | It is soluble in water (50mg/ml,) |
| Formula Weight | 183.94 |
| Concentration or Composition (by Analyte or Components) | 100mM aq. soln. |
| Odor | Odorless |
| Chemical Name or Material | Sodium orthovanadate |
Safety and Handling
| Hazard Category | H335 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only