missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside, For Nitrogen (Ammonia), Nitrogen (Organic), 0.05% (w/v), LabChem™

Catalog No. LC247002
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC247002 1 L
LC247001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC247002 Supplier LabChem Supplier No. LC247002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13755-38-9 , 7732-18-5
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Tan
CAS Max % 99.95, 0.05
Linear Formula Na2Fe(CN)5(NO)·2H2O
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 297.25
Percent Purity 0.05% (w/v)
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium Nitroprusside, For Nitrogen (Ammonia), Nitrogen (Organic)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature