missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitroprusside, 0.5 g/L, Ricca Chemical
$95.83 - $165.57
Chemical Identifiers
| CAS | 13755-38-9 |
|---|---|
| Molecular Formula | C5H4FeN6Na2O3 |
| Molecular Weight (g/mol) | 297.95 |
| MDL Number | MFCD00149192 |
| InChI Key | JFPDVRGGFBUORC-UHFFFAOYSA-N |
| Synonym | nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o |
| PubChem CID | 11953895 |
| IUPAC Name | disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate |
| SMILES | O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
Chemical Identifiers
| 13755-38-9 | |
| 297.95 | |
| JFPDVRGGFBUORC-UHFFFAOYSA-N | |
| 11953895 | |
| O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
| C5H4FeN6Na2O3 | |
| MFCD00149192 | |
| nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
| disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate |
Specifications
| Golden-Tan | |
| 98.0,0.05 | |
| C5H4FeN6Na2O3 | |
| nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
| O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] | |
| 297.95 | |
| Laboratory | |
| Aqueous Solution | |
| Liquid | |
| Sodium Nitroprusside |
| 13755-38-9 , 7732-18-5 | |
| 100.0,0.05 | |
| MFCD00149192 | |
| JFPDVRGGFBUORC-UHFFFAOYSA-N | |
| disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate | |
| 11953895 | |
| Amber Glass Bottle | |
| 500 mL | |
| 0.5g/L |