missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside, 0.5 g/L, Ricca Chemical

Catalog No. 749832
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
749832 1 L Amber Glass Bottle
749816 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 749832 Supplier Ricca Chemical Company Supplier No. 749832

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13755-38-9 , 7732-18-5
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Golden-Tan
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.5g/L
Chemical Name or Material Sodium Nitroprusside