missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroferricyanide Dihydrate (Crystalline/Certified ACS), Fisher Chemical

Catalog No. S350100
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S350100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S350100 Supplier Thermo Fisher Scientific Supplier No. S350 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Red
pH 5
Physical Form Crystalline Solid
Quantity 100 g
Identification Pass Test
Packaging Glass Bottle
Assay Percent Range 99.0 to 102.0 %
Density 1.720g/cm³
Linear Formula Na2Fe(CN)5(NO)·2H2O
Formula Weight 297.95
Percent Purity 99.0 to 102.0%
Grade Certified ACS
Chemical Name or Material Sodium Nitroferricyanide Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Toxic by inhalation, in contact with skin and if swallowed. May cause pulmonary edema. May cause central nervous system effects. Contact with acids liberates very toxic gas. Hygroscopic. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe vapors/dust. Do not ingest. Keep away from acids. Keep away from acids. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:1
Instability:1