missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrite, 99.999% (metals basis)

Catalog No. AA4422722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4422722 100 g
AA4422714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4422722 Supplier Thermo Scientific Chemicals Supplier No. 04422722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7632-00-0 ∣ NNaO2 ∣ 69.00 g/mol

Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor and in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Specifications

Melting Point 271°C
Color White
Physical Form Crystals
Quantity 100 g
Odor Odorless
Sensitivity Hygroscopic
Assay Percent Range 99.999% (metals basis)
Density 2.17
Solubility Information Soluble in water,methanol,ethanol,ammonia,nitric acid. Slightly soluble diethyl ether.
Formula Weight 69
Percent Purity 100%
Grade Ungraded
Chemical Name or Material Sodium nitrite
Show More Show Less
Hazard Category H272-H301-H319-H373
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 231-555-9
RTECSNumber RA1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only