missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, 99.0% min., ACS, MilliporeSigma™

Catalog No. MSX06657
Change view
Click to view available options
Quantity:
500 g
12 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX06657 12 kg
MSX06651 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MSX06657 Supplier Technidata Supplier No. SX06657

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Color White or Colorless
Physical Form Crystals or Powder
Quantity 12 kg
Assay Percent Range 99.0% min.
Linear Formula NaNO2
Formula Weight 69
Grade ACS/EP/Reagent
Ignition Point 510°C
Density 2.1g/cm3 (20°C); Bulk Density: 1200 kg/m3
Chemical Name or Material Sodium Nitrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions